Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC2/3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$181.90 - $487.50

Specifications

Antigen VDAC2/3
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056463T
SDP
View Documents
Novus Biologicals
NB10056463SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056463
SDP
View Documents
Novus Biologicals
NB10056463
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

VDAC2/3 Polyclonal specifically detects VDAC2/3 in Human, Bovine, Canine, Feline, Primate samples. It is validated for Western Blot.
Specifications

Specifications

VDAC2/3
Western Blot 1-3 ug/ml
Polyclonal
Purified
RUO
PBS containing 0.05% BSA with 0.05% Sodium Azide
FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2
VDAC2
IgG
Protein G purified
Reacts with Bovine, Cat, Chimpanzee, Dog and Human. 92% sequence identity with Mouse, Rat, Chicken and Zebrafish proteins.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Human, Bovine, Canine, Feline, Primate
P45880
7417
Amino acids 120-132 (KKSGKIKSSYKRE) of human VDAC2 protein were used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.