Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10074560 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074560 Supplier Novus Biologicals Supplier No. NB10074560

Chicken Polyclonal Antibody

VDAC2 Polyclonal specifically detects VDAC2 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA.

Specifications

Antigen VDAC2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100 - 1:2000
Formulation PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2
Gene Symbols VDAC2
Host Species Chicken
Immunogen VDAC2 Protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7417
Test Specificity This detects VDAC2 in mouse, rat, human, rabbit, and bovine samples.
Target Species Human, Mouse, Rat, Bovine, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.