Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDR/NR1I1/Vitamin D Receptor Antibody (3C7), Novus Biologicals™
SDP

Catalog No. NB298467 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298467 100 μL
NB298466 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298467 Supplier Novus Biologicals Supplier No. NBP326255100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VDR/NR1I1/Vitamin D Receptor Monoclonal antibody specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen VDR/NR1I1/Vitamin D Receptor
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 3C7
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor
Host Species Rabbit
Immunogen A synthesized peptide derived from Human VDR/NR1I1/Vitamin D Receptor [UniProt P11473]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7421
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.