Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDR/NR1I1/Vitamin D Receptor Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB055933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB055933 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB055933 Supplier Novus Biologicals Supplier No. NBP298840G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VDR/NR1I1/Vitamin D Receptor Polyclonal specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VDR/NR1I1/Vitamin D Receptor
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human VDR/NR1I1/Vitamin D Receptor.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7421
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.