Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VE-Cadherin Antibody (048), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB099480 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB099480 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB099480 Supplier Novus Biologicals Supplier No. NBP289498AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection.

Specifications

Antigen VE-Cadherin
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 48
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human VE-Cadherin (CAA56306.1; Met1-Gln593).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1003
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.