Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VE-Cadherin Antibody (162709) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB402652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402652 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402652 Supplier Novus Biologicals Supplier No. FAB1002Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

VE-Cadherin Monoclonal antibody specifically detects VE-Cadherin in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen VE-Cadherin
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 162709
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse VE-Cadherin, Asp46-Gln592 (Gly67 del, Ile69Asp, Lys70Gln), Accession # 2208309A
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1003
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.