Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VE-Cadherin Antibody (16B1), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N20342PE594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N20342PE594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N20342PE594 Supplier Novus Biologicals Supplier No. NBP200342PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen VE-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone 16B1
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin
Gene Symbols CDH5
Host Species Mouse
Immunogen The immunogen for this antibody is VE Cadherin.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1003.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.