Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VE-Statin/EGFL7 Antibody (OTI2B5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP274846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274846 Supplier Novus Biologicals Supplier No. NBP274846
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VE-Statin/EGFL7 Monoclonal specifically detects VE-Statin/EGFL7 in Human samples. It is validated for Western Blot.

Specifications

Antigen VE-Statin/EGFL7
Applications Western Blot
Classification Monoclonal
Clone OTI2B5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VE-STATIN, Zneu1
Gene Symbols EGFL7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 24-273 of human EGFL7(NP_057299) produced in E.coli.
Molecular Weight of Antigen 29.4 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 51162
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.