Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF 121 Antibody (064), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB354603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354603 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354603 Supplier Novus Biologicals Supplier No. NBP289193APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VEGF 121 Monoclonal antibody specifically detects VEGF 121 in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen VEGF 121
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 064
Conjugate APC
Dilution ELISA, Sandwich ELISA Detection
Formulation PBS
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF 121 (Uniprot#: P15692-9; Met1-Arg147).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7422
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.