Learn More
Description
Specifications
Specifications
| Antigen | VEGF 121 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 64 |
| Conjugate | DyLight 488 |
| Dilution | ELISA, Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF 121 (P15692-9; Met1-Arg147). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
