Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB1002381X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002381X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002381X Supplier Novus Biologicals Supplier No. NB1002381AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VEGF Polyclonal specifically detects VEGF in Bovine, Canine, Chicken, Equine, Guinea Pig, Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.

Specifications

Antigen VEGF
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P15692
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human VEGFA protein sequence (between residues 180-232). [Swiss-Prot: P15692]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, GPCR, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity Human, mouse, rat, horse, chicken, guinea pig, dog, and cow. Predicted to react with sheep and primate based on 100% sequence homology.
Target Species Bovine, Canine, Chicken, Equine, Guinea Pig, Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.