Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Antibody (VEGF/1063), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB362542 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362542 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362542 Supplier Novus Biologicals Supplier No. NBP311437IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGF Monoclonal antibody specifically detects VEGF in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen VEGF
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone VEGF/1063
Conjugate DyLight 755
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Host Species Mouse
Immunogen Human VEGF recombinant protein (Uniprot: P15692)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 7422
Target Species Human, Mouse, Rat, Canine, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.