Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Antibody (VG76e) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100648 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100648 0.1 mL
NB100648SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100648 Supplier Novus Biologicals Supplier No. NB100648
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 13 publications

VEGF Monoclonal specifically detects VEGF in Bovine, Goat, Human, Pig, Rat, Sheep samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot.

Specifications

Antigen VEGF
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone VG76e
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Radioimmunoassay reported in scientific literature
Gene Accession No. P15692
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Mouse
Immunogen Recombinant human VEGF189 expressed in E.coli [UniProt# P15692]
Molecular Weight of Antigen 40 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity Using recombinant proteins, it was demonstrated that VG76e recognizes the 121, 165, and 189 isoforms of VEGF
Target Species Bovine, Rat, Goat, Human, Pig, Rat, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.