Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Antibody (VG76e) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB001483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001483 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001483 Supplier Novus Biologicals Supplier No. NBP2625640.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody has been used in 1 publication

VEGF Monoclonal specifically detects VEGF in Human, Mouse, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen VEGF
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone VG76e
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Formulation PBS
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Rabbit
Immunogen Human VEGF189 expressed in E. coli.
Quantity 0.025 mg
Gene ID (Entrez) 7422
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.