Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-B Antibody (RM0008-6E72), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1061024A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1061024A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1061024A30 Supplier Novus Biologicals Supplier No. NB11061024AF350
Add to Cart
Add to Cart

Rat Monoclonal Antibody

VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VEGF-B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0008-6E72
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL
Gene Symbols VEGFB
Host Species Rat
Immunogen Mouse recombinant VEGF-B
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7423
Test Specificity This antibody recognizes mouse VEGF-B 186 but not 167
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.