Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-B Antibody (RM0008-6E72) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018002 0.025 mg
NB11061024 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018002 Supplier Novus Biologicals Supplier No. NB110610240.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VEGF-B
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone RM0008-6E72
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:400-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL
Gene Symbols VEGFB
Host Species Rat
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse VEGF-B (186).
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7423
Test Specificity VEGFB (RM0008-6E72)
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.