Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-B Antibody (SR1236), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32221650UL
Description
VEGF-B Monoclonal antibody specifically detects VEGF-B in Human, Mouse samples. It is validated for Western BlotSpecifications
VEGF-B | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
SR1236 | |
Western Blot 1:500-1:2000 | |
vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
A synthesized peptide derived from human VEGF-B (Uniprot #: P49765) | |
50 μg | |
Angiogenesis, Cell Cycle and Replication | |
7423 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction