Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-B Antibody (SR1236), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | VEGF-B |
---|---|
Clone | SR1236 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
VEGF-B Monoclonal antibody specifically detects VEGF-B in Human, Mouse samples. It is validated for Western BlotSpecifications
VEGF-B | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
A synthesized peptide derived from human VEGF-B (Uniprot #: P49765) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1236 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cell Cycle and Replication | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7423 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title