Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-C Antibody (107/F10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP118626
Description
VEGF-C Monoclonal antibody specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
| VEGF-C | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 - 5 ug/mL, Immunohistochemistry 2 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 2 - 10 ug/mL | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody detects human and rat VEGF-C in Western blots. No cross-reactivity is shown with VEGF-A. | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 107/F10 | |
| Unconjugated | |
| PBS with No Preservative | |
| VEGFC | |
| Recombinant human VEGF-C | |
| 0.1 mg | |
| Angiogenesis, Cancer | |
| 7424 | |
| Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. Please note the sample size of this product will be provided in reconstituted liquid form. | |
| Store at -80C in the dark. Avoid freeze-thaw cycles. | |
| IgG2 α |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction