Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-C Antibody (107/F10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$214.28 - $579.29
Specifications
| Antigen | VEGF-C |
|---|---|
| Clone | 107/F10 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
VEGF-C Monoclonal antibody specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
| VEGF-C | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer | |
| 7424 | |
| Recombinant human VEGF-C | |
| Primary | |
| Store at -80C in the dark. Avoid freeze-thaw cycles. |
| 107/F10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
| VEGFC | |
| IgG2 α | |
| Protein G purified | |
| This antibody detects human and rat VEGF-C in Western blots. No cross-reactivity is shown with VEGF-A. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title