Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-C Antibody (MM0006-2E65) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB017977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017977 0.025 mg
NB11060971 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017977 Supplier Novus Biologicals Supplier No. NB110609710.025MG

Mouse Monoclonal Antibody

VEGF-C Monoclonal specifically detects VEGF-C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VEGF-C
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone MM0006-2E65
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:25-50, Immunohistochemistry-Paraffin 1:25-1:50
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P49767
Gene Alias Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Gene Symbols VEGFC
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with recombinant human VEGF-C recombinant protein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7424
Test Specificity No cross-reactivity is shown with human VEGFA, VEGF-B and VEGF-D.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.