Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-C Antibody (OTI4A1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274845V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274845V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274845V Supplier Novus Biologicals Supplier No. NBP274845V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGF-C Monoclonal specifically detects VEGF-C in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen VEGF-C
Applications Western Blot
Classification Monoclonal
Clone OTI4A1
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Gene Symbols VEGFC
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 112-227 of human VEGF-C (NP_005420) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7424.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.