Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-D Antibody (MM0007-7E79), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N1060973A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1060973A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1060973A94 Supplier Novus Biologicals Supplier No. NB11060973AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VEGF-D
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0007-7E79
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor
Gene Symbols VEGFD
Host Species Mouse
Immunogen Human recombinant VEGF-D
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2277
Test Specificity VEGFD (MM0007-7E79). No cross-reactivity is shown with VEGF and VEGF-B.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.