Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-D Antibody (MM0007-7E79), PE, Novus Biologicals™
SDP

Catalog No. NB344871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344871 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344871 Supplier Novus Biologicals Supplier No. NB11060973PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGF-D Monoclonal antibody specifically detects VEGF-D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen VEGF-D
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0007-7E79
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor
Host Species Mouse
Immunogen Human recombinant VEGF-D
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 2277
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.