Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR-1/FLT-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7206240 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8936 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8936 Supplier Proteintech Group Inc Supplier No. 136871AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VEGFR 1 is a cell membrane receptor kinase. It has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.

Specifications

Antigen VEGFR-1/FLT-1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FLT1
Gene Accession No. P17948, P35969, P53767
Gene Alias FLT, FLT 1, FLT1, FLT-1, Fms like tyrosine kinase 1, FRT, sflt1, Tyrosine protein kinase FRT, VEGF receptor 1, VEGFR 1, VEGFR1, VEGFR-1
Gene Symbols Flt1
Host Species Rabbit
Immunogen VEGFR-1/FLT-1 Fusion Protein Ag4558
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14254, 2321, 54251
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.