Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR1/Flt-1 Antibody (MAB0702), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB221963647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB221963647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB221963647 Supplier Novus Biologicals Supplier No. NBP221963AF647

Mouse Monoclonal Antibody

VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen VEGFR1/Flt-1
Applications Western Blot
Classification Monoclonal
Clone MAB0702
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Gene Symbols FLT1
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified extracellular domain of recombinant human VEGFR1. VEGFR-1 ( Antagonistic) EC domain, AAC50060.
Purification Method Affinity Purified
Quantity 100 μL
Research Discipline Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 2321
Test Specificity This antibody recognizes both human and mouse VEGFR1 in western blot. It shows no cross-reactivity with human or mouse VEGFR2.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.