Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR1/Flt-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100527 0.1 mL
NB100527SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100527 Supplier Novus Biologicals Supplier No. NB100527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

VEGFR1/Flt-1 Polyclonal specifically detects VEGFR1/Flt-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen VEGFR1/Flt-1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:250 - 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:250 - 1:500
Formulation Tris-Glycine, 0.15 M NaCl with 0.05% Sodium Azide
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Gene Symbols FLT1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the mouse VEGF Receptor 1 protein (between residues 800-900) [Uniprot: P35969]. There is 87% identity between the immunogen used for this production and the VEGF Receptor 2 protein.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 2321
Test Specificity This antibody targets VEGFR-1 but has significant cross-reactivity with the VEGFR-2 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.