Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR1/Flt-1 Antibody (Z9P4F3*H10), FITC, Novus Biologicals™
SDP

Catalog No. NBP250408F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250408F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250408F Supplier Novus Biologicals Supplier No. NBP250408F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Zebrafish samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen VEGFR1/Flt-1
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Z9P4F3*H10
Conjugate FITC
Dilution ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Gene Symbols FLT1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KEIPDDEQSD (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 2321
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.