Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR2/KDR/Flk-1 Antibody (20I6) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018381 0.02 mg
NBP118646 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018381 Supplier Novus Biologicals Supplier No. NBP1186460.02MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen VEGFR2/KDR/Flk-1
Applications Immunofluorescence, Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry, CyTOF, Immunohistochemistry, ELISA, Western Blot
Classification Monoclonal
Clone 20I6
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, Flow Cytometry 2-5 ug/mL, ELISA 1-10 ug/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Frozen 6-30 ug/mL, CyTOF-ready
Formulation No buffer with No Preservative
Gene Alias CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2
Gene Symbols KDR
Host Species Mouse
Immunogen Recombinant human soluble KDR (D7)
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3791
Test Specificity The monoclonal antibody will detect native human VEGFR-2/KDR in ELISA experiments and on the surface of different human cell types.
Reconstitution Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.