Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR2/KDR/Flk-1 Antibody (89106) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB372060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372060 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372060 Supplier Novus Biologicals Supplier No. FAB3572Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for CyTOF, Flow Cytometry, Neutralization.

Specifications

Antigen VEGFR2/KDR/Flk-1
Applications CyTOF, Flow Cytometry, Neutralization
Classification Monoclonal
Clone 89106
Conjugate DyLight 680
Dilution Flow Cytometry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR2/KDR/Flk-1, Ala20-Glu764, Accession # P35968
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3791
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.