Learn More
VEGFR2/KDR/Flk-1 Antibody (Avas12a1), Allophycocyanin, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | VEGFR2/KDR/Flk-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Avas12a1 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Alias | CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2 |
| Gene Symbols | KDR |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was Flk1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.