Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR2/KDR/Flk-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100529 0.1 mL
NB100529SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100529 Supplier Novus Biologicals Supplier No. NB100529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VEGFR2/KDR/Flk-1 Polyclonal specifically detects VEGFR2/KDR/Flk-1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen VEGFR2/KDR/Flk-1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250 - 1:1000, Immunocytochemistry/Immunofluorescence 1:50 - 1:500
Formulation Tris-Glycine, 0.15 M NaCl with 0.05% Sodium Azide
Gene Alias CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, FLK-1, FLK1tyrosine kinase growth factor receptor, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGFR, VEGFR2, VEGFR-2
Gene Symbols KDR
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the mouse VEGF Receptor 2 protein (between residues 1200-1300). [Swiss-Prot: P35918]
Molecular Weight of Antigen 150 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3791
Test Specificity Human and mouse. Immunogen sequence has 100% homology to rat.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.