Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Velvet pad, 6 Inch Square (Pack), Thick Velvet Pad
Click to view available options
Quantity:
10/Pk.
Description
- Velvet Pad is a consumable that is used with the Replica Plating Apparatus and allows of colony transfer from Agar Plate while retaining the colony orientation.
- Velvet pad is used in Replica plating technique. This technique used for screening auxotrophy and antibiotic resistance.

Specifications
Specifications
Content And Storage | Room Temperature (15°C to 30°C) |
Description | Velvet Pad is a consumable that is used with the Replica Plating Apparatus and allows of colony transfer from Agar Plate while retaining the colony orientation. |
For Use With (Application) | Velvet pad is used in Replica plating technique. This technique used for screening auxotrophy and antibiotic resistance. |
Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction