Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Venezuelan Equine Encephalitis Virus NSP1 Antibody (HL1472) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294881 Supplier Novus Biologicals Supplier No. NBP325736
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Venezuelan Equine Encephalitis Virus NSP1 Monoclonal antibody specifically detects Venezuelan Equine Encephalitis Virus NSP1 in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Venezuelan Equine Encephalitis Virus NSP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL1472
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias mRNA-capping enzyme nsp1, VEEV Non-Structural Protein 1, VEEV NSP1, Venezuelan Equine Encephalitis Virus Non-Structural Protein 1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-term region of Venezuelan Equine Encephalitis Virus NSP1. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.