Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Venlafaxine hydrochloride
SDP

Catalog No. 291750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-175-0 50 mg
29-171-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-175-0 Supplier R&D Systems™ Supplier No. 2917/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-HT and NA reuptake inhibitor (SNRI)

Dual serotonin/noradrenalin re-uptake inhibitor that displays ∽ 30-fold higher affinity for SERT than NET (Ki values are 82 and 2480 nM respectively). Antidepressant; increases swimming and climbing behavior in the forced-swim test in rats.

Chemical Identifiers

CAS 99300-78-4
Molecular Formula C17H28ClNO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 313.86
Purity >99%
Molecular Weight (g/mol) 313.866
Chemical Name or Material Venlafaxine hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.