Learn More
Restek™ Veriflo Flow Controllers
SDP
Available in Siltek-treated and stainless steel versions.
$1147.00 - $1378.00
Specifications
Type Veriflo SC423 XL Flow Controller
View More Specs
Products 8
Catalog Number Mfr. No. Material Size (Metric) Orifice Price Quantity & Availability  
Catalog Number Mfr. No. Material Size (Metric) Orifice Price Quantity & Availability  
06-712-264
SDP
View Documents
Restek™
24264
Stainless Steel 0.015 cm
Each for $1,170.00
Only null left
Add to Cart
06-712-260
SDP
View Documents
Restek™
24260
Stainless Steel 0.003 cm
Each for $1,200.00
Only null left
Add to Cart
06-712-261
SDP
View Documents
Restek™
24261
Stainless Steel 0.004 cm
Each for $1,170.00
Only null left
Add to Cart
06-712-248
SDP
View Documents
Restek™
24238
Siltek Treated Without orifice
Each for $1,318.00
Only null left
Add to Cart
06-712-263
SDP
View Documents
Restek™
24263
Stainless Steel 0.007 cm
Each for $1,170.00
Only null left
Add to Cart
06-712-262
SDP
View Documents
Restek™
24262
Stainless Steel 0.005 cm
Each for $1,170.00
Only null left
Add to Cart
06-712-249
SDP
View Documents
Restek™
24239
Stainless Steel Without orifice
Each for $1,147.00
Only null left
Add to Cart
06-712-255
SDP
View Documents
Restek™
24255
Siltek Treated 0.003 cm
Each for $1,378.00
Only null left
Add to Cart
Description
Description
  • Available with or without a critical orifice
  • Interchangeable critical orifice
Order Info
Order orifices for alternate sampling times, or replacement orifices, separately.
Specifications
Specifications
Veriflo SC423 XL Flow Controller
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.