Learn More
Vernier Vernier Chemical Polarimeter
SDPSCI_ED
Catalog No. s16121nd Shop All Vernier Software Products
Click to view available options
:
Chemical Polarimeter
Catalog No. S16121ND
Supplier: Vernier CHEMPOL
Includes
Chemical Polarimeter, One Chemical Polarimeter Sample Cell, User manual
Used for measuring rotation of plane-polarized light caused by optically active substances such as organic, inorganic, or biological compounds.
  • Used to measure chiral properties of optically active samples without chemically modifying or destroying sample
  • Is vertical polarimeter that uses 589nm LED, fixed polarizer, and manually rotated polarizer to detect changes in rotation of plane-polarized light in presence of optically active compound
  • Compatible with LabQuest 2, Original LabQuest, LabQuest Mini, SensorDAQ, LabPro, TI-Nspire Lab Cradle interfaces
  • Optical Encoder: Bidirectional, quadrature encoder, 360 cycle per revolution
Specifications
Description Chemical Polarimeter
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.