Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Lanyard
SDPSCI_ED

Catalog No. s16188nd Shop All Vernier Software Products
Click to view available options
:
Lanyard
Works as a LabQuest neck strap to prevent accidental drops while doing field studies Can also be attached to a calculator cradle and used with a LabPro or CBL 2

Specifications

Color Black
For Use With (Equipment) LabQuest, calculator cradle, LabPro or CBL 2
For Use With (Application) Securing instruments
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.