Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Versaflo™ Respiratory Faceshield Assembly
Faceshield assembly is a lightweight, comfortable PAPR headtop that combines a respirator and faceshield, with the option of adding earmuffs for hearing protection.
$521.11 - $606.73
Specifications
Color | Gray |
---|---|
Product Type | Faceshield Assembly |
For Use With (Equipment) | 3M™ Versaflo™ PAPR and Supplied Air Systems |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
19-039-900
|
3M™
7100086942 |
Comfort Faceseal |
Case for $521.11
|
|
|||||
19-039-901
|
3M™
7100086969 |
Flame Resistant Faceseal |
Case for $606.73
|
|
|||||
Description
- Complete, fully assembled PAPR Headtop allows for use right out of the box
- Flexible application - Use with any 3M Versaflo system
- Hearing protection compatible with M-985 earmuffs
- Comfortable PAPR alternative to negative pressure respirators and standard faceshields
- Meets the requirements of the ANSI Z87.1-2010 standard for limited eye and face protection devices
- Easy-to-use design with ratchet suspension and liftable visor
- Popular in light industry where respirators and face protection are typically used
- Combine the M-206 with 3M Versaflo PAPR or Supplied Air components to create a complete respiratory assembly.
Specifications
Gray | |
Faceshield Assembly | |
3M™ Versaflo™ PAPR and Supplied Air Systems | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title