Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Versican Antibody (S351-23), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB222408700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222408700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222408700 Supplier Novus Biologicals Supplier No. NBP222408AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Versican Monoclonal specifically detects Versican in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Versican
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S351-23
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Chondroitin sulfate proteoglycan 2WGN, Chondroitin sulfate proteoglycan core protein 2, CSPG2DKFZp686K06110, ERVR, GHAP, Glial hyaluronate-binding protein, Large fibroblast proteoglycan, PG-MWGN1, versican, versican core protein, versican proteoglycan
Gene Symbols VCAN
Host Species Mouse
Immunogen Fusion protein amino acids 362-585 (glycosaminoglycan alpha domain) of mouse Versican core protein
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1462
Test Specificity Detects >350kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.