Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Versican Rabbit anti-Human, Mouse, Rat, Clone: 6D8K9, Novus Biologicals™
SDP

Catalog No. NB166777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166777 100 μg
NB166778 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166777 Supplier Novus Biologicals Supplier No. NBP315841100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Versican Monoclonal antibody specifically detects Versican in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Versican
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6D8K9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Chondroitin sulfate proteoglycan 2WGN, Chondroitin sulfate proteoglycan core protein 2, CSPG2DKFZp686K06110, ERVR, GHAP, Glial hyaluronate-binding protein, Large fibroblast proteoglycan, PG-MWGN1, versican, versican core protein, versican proteoglycan
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 3200-3396 of human Versican (P13611). QGAHLTSILSHEEQMFVNRVGHDYQWIGLNDKMFEHDFRWTDGSTLQYENWRPNQPDSFFSAGEDCVVIIWHENGQWNDVPCNYHLTYTCKKGTVACGQPPVVENAKTFGKMKPRYEINSLIRYHCKDGFIQRHLPTIRCLGNGRWAIPKITCMNPSAYQRTYSMKYFKNSSSAKDNSINTSKHDHRWSRRWQESRR
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1462
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.