Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Verso 1-step RT-qPCR Mix, low ROX

Catalog No. AB4102A
Change view
Click to view available options
No. of Reactions:
200 Reactions
400 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
AB4102A 200 Reactions
AB4102C 400 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB4102A Supplier Thermo Scientific™ Supplier No. AB4102A
Add to Cart
Add to Cart

The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures.

The Thermo Scientific Verso 1-step RT-qPCR Low ROX Mix Kit for probe chemistry offers a quick and simple way to detect mRNA.

An optional RT Enhancer is also provided and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment. All Verso 1-step RT-qPCR kits incorporate an inert dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

In a 1-step approach to RT-qPCR, both the RT step and qPCR steps are performed in a single tube (1-step RT-qPCR). Adopting a 1-step approach is ideal when no cDNA storage is necessary for subsequent experiments. The approach minimizes the risk of contamination and improves reproducibility by reducing sample handling.

  • Eliminate the need for DNase treatments - RT Enhancer (optional) prevents gDNA carry-over
  • RT and qPCR steps performed in single tube - convenience and reduced contamination risk
  • Use of the entire RT reaction as template for qPCR - improved sensitivity
  • Verso RT enzyme is highly sensitive with broad dynamic range - accurate detection of RNA from 1pg to 1μg
  • Verso RT enzyme has a wide working temperature range (42 to 57°C) - improved success with GC-rich and other difficult templates
  • Contains an inert blue dye for ease of pipetting during reaction set up
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
•1 x 50 μL Verso Enzyme Mix
•1 x 250μL RT enhancer
•2 x 1.25 mL 2X 1-Step qPCR Low ROX Mix

Store at -20°C
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR
For Use With (Equipment) 7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
Product Line Verso
Product Type 1-Step RT-qPCR Mix
Quantity 200 reactions
Sufficient For 200 Reactions
For Use With (Application) Gene Expression, Pathogen Detection
No. of Reactions 200 Reactions
Sample Type RNA
Show More Show Less
What is the RT Enhancer that is included in all Verso kits?

The RT Enhancer is a unique enzyme that degrades contaminating genomic DNA at the start of the RT step. It is equally as effective as DNase I treatment but more convenient as it eliminates the need for a separate DNase I incubation prior to RT-PCR.

Do I need to treat my RNA with DNase I before running RT-qPCR with Verso cDNA Synthesis Kit?

No, you do not need to treat your RNA with DNase I before running RT-qPCR with the Verso cDNA Synthesis Kit. The RT enhancer included in the Verso kit degrades any contaminating double-stranded DNA during the reverse transcription process, eliminating the need for a separate DNase I treatment.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.