Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Vertical Electrophoresis Systems
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W x H):
6.7 x 5.1 x 5.5 in.
9.8 x 5.1 x 7.7 in.
Gel Dimensions:
10 x 10 cm
16 x 14 cm
Volume (Metric):
300 mL
550 mL
Description
Key Features
- Systems are double-sided, allowing two protein gels to be run simultaneously
- Gel assemblies don't need to be moved after the gel is set
- Sliding clamp assemblies make system setup a snap and prevent leaks
- Electrode configuration reduces run times
- Increased electrode surface area provides exceptional field uniformity
Safety Features
- Asymmetrical shape prevents mismatching of electrodes
- Interlocking safety lid with attached power cords protect against electric shock

Specifications
Specifications
Gel Dimensions | 16 x 14 cm |
Includes | Upper buffer chamber with cooling chamber, lower buffer chamber, safety lid (with attached power supply leads), gaskets, clamp assemblies, gel caster, four blank and two notched glass plates, two 10-well and two 15-well, 1.5mm thick combs, eight, 1.5mm thick side spacers |
Volume (English) Buffer | 18.59 oz. |
Volume (Metric) | 550 mL |
Volume (Metric) Buffer | 550 mL |
Warranty | 3 Years |
Dimensions (L x W x H) | 9.8 x 5.1 x 7.7 in. |
Length (English) | 9.8 in. |
Length (English) Gel | 5.5 to 6.29 in. |
Length (Metric) | 24.89 cm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction