Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vesicular Stomatitis Indiana Virus Glycoprotein Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB293662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293662 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293662 Supplier Novus Biologicals Supplier No. NBP326070JF549

Rabbit Polyclonal Antibody

Vesicular Stomatitis Indiana Virus Glycoprotein Polyclonal antibody specifically detects Vesicular Stomatitis Indiana Virus Glycoprotein in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Vesicular Stomatitis Indiana Virus Glycoprotein
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Vesicular Stomatitis Indiana Virus Glycoprotein (Accession#: NP_041715.1, Uniprot#: P03522, Met1-Ser467)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.