Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vFLIP Antibody (6/14), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB308478 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308478 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308478 Supplier Novus Biologicals Supplier No. NBP278138MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

vFLIP Monoclonal antibody specifically detects vFLIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen vFLIP
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2024-06-14 00:00:00
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Full-length recombinant vFLIP expressed in Escherichia coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.