Learn More
Description
Specifications
Specifications
| Antigen | VG5Q |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA reported in scientific literature, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin |
| Formulation | Whole antisera with No Preservative |
| Gene Accession No. | Q8N302 |
| Gene Alias | Angiogenic factor VG5Q, angiogenic factor with G patch and FHA domains 1, FLJ10283hVG5Q, GPATC7G patch domain-containing protein 7, GPATCH7HUS84971, HSU84971, VG5QVasculogenesis gene on 5q protein |
| Gene Symbols | AGGF1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
