Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VG5Q Antibody, Novus Biologicals™
SDP

Catalog No. NB100454SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100454SS 0.025 mL
NB100454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100454SS Supplier Novus Biologicals Supplier No. NB100454SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

VG5Q Polyclonal specifically detects VG5Q in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen VG5Q
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA reported in scientific literature, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin
Formulation Whole antisera with No Preservative
Gene Accession No. Q8N302
Gene Alias Angiogenic factor VG5Q, angiogenic factor with G patch and FHA domains 1, FLJ10283hVG5Q, GPATC7G patch domain-containing protein 7, GPATCH7HUS84971, HSU84971, VG5QVasculogenesis gene on 5q protein
Gene Symbols AGGF1
Host Species Rabbit
Immunogen A synthetic peptide mapping to an epitope within residues 500-600 of the human VG5Q protein. [UniProt# Q8N302]
Molecular Weight of Antigen 100 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 55109
Test Specificity This is specific for human VG5Q.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.