Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGCNL1 Antibody (S187-7), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP222410IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222410IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222410IR Supplier Novus Biologicals Supplier No. NBP222410IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VGCNL1 Monoclonal specifically detects VGCNL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen VGCNL1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S187-7
Conjugate DyLight 755
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA430M15.1, CanIonFLJ44659, FLJ23913, FLJ44764, four repeat voltage-gated ion channel, MGC74524, sodium leak channel non-selective protein, sodium leak channel, non-selective, VGCNL1, voltage gated channel like 1, Voltage gated channel-like protein 1
Gene Symbols NALCN
Host Species Mouse
Immunogen Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 259232
Test Specificity Detects approx 200kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.