Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGLUT1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100183705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100183705 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100183705 Supplier Novus Biologicals Supplier No. NB10018370.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

VGLUT1 Polyclonal specifically detects VGLUT1 in Canine, Mouse, Rat samples. It is validated for Electron Microscopy, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot.

Specifications

Antigen VGLUT1
Applications Electron Microscopy, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Radioimmunoassay 1:500, Electron Microscopy 1:200
Formulation Tris 0,1M, glycine 0,1M, sucrose 2%. with No Preservative
Gene Accession No. Q62634
Gene Alias BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1
Gene Symbols SLC17A7
Host Species Rabbit
Immunogen A 17 residue synthetic peptide derived from C-terminal domain of rat VGLUT1.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57030
Test Specificity Reacts with rat 55-60 kDa VGLUT1 protein of BON cells stably expressing VGLUT1 and of total rat brain extracts. No cross reactivity was detected with VGLUT2 and VGLUT3
Reconstitution Reconstitute with deionized water.
Target Species Canine, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.