Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGLUT1 Antibody (S28-9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259329
Description
VGLUT1 Monoclonal specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| VGLUT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| SLC17A7 | |
| Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1 | |
| 100 μg | |
| Primary | |
| Detects 52kDa. No cross-reactivity against VGlut2. | |
| Store at -20C. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| S28-9 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 57030 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction